missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPKAPK3 Polyclonal Antibody
MAPKAPK3 Polyclonal Antibody
Brand: Invitrogen PA517270
This item is not returnable.
View return policy
Description
MAPKAPK3 Polyclonal Antibody for Western Blot
Specifications
| MAPKAPK3 | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q16644, Q66H84 | |
| MAPKAPK3 | |
| Synthetic peptide corresponding to the amino-terminal region of human MAPKAPK-3. | |
| 100 μL | |
| Primary | |
| Human, Rat, Monkey | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| MAPKAPK3 | |
| 3PK; AI874665; chromosome 3p kinase; MAP kinase-activated protein kinase 3; MAPK-activated protein kinase 3; MAPKAP kinase 3; MAPKAP3; MAPKAPK3; MAPKAP-K3; MAPKAPK-3; MapkKapk3; mitogen activated protein kinase-activated protein kinase 3; mitogen-activated protein kinase-activated protein kinase 3; MK3; MK-3; OTTHUMP00000210806; OTTHUMP00000210807 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 315994, 7867 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction