Learn More
Hamilton™ Manual HPLC Cemented Needle Syringes
Click to view available options
Volume (Metric):
250 μL
Connection Type:
Cemented
Luer Tip, Cemented
Unit Size:
Each
Description
- Needle is blunt and electro-polished to slide smoothly into the injection port
- Available as standard cemented needle or Luer tip cemented needle
- 51mm in length with point style 3
Specifications
Specifications
| Needle Gauge | 22s G |
| Volume (Metric) | 250 μL |
| Length (Metric) Needle | 51 mm |
| Needle Point Style | 3 |
| Connection Type | Luer Tip, Cemented |