missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Mannitol Salt Agar (Chapman Agar)
Description
- BAM/EP/HP/ISO/USP
- Selective medium for the isolation of pathogenic staphylococci
- According to the Pharmacopoeial Harmonized Methodology and the ISO 22718 standard.
Specifications
Specifications
| Certifications/Compliance | ISO 22718 |
| Form | Solid |
| Media Type | Mannitol Salt Agar |
| Product Type | Selective Medium |
| Target Organism Class | Gram-positive Organisms |
| Formulation | Beef Extract 1.00 g/L, Pancreatic Digest of Casein 5.00 g/L, Peptic Digest of Meat 5.00 g/L, Sodium Chloride 75.00 g/L, D-Mannitol 10.00 g/L, Phenol Red 0.025 g/L, Agar 15.00 g/L |
| For Use With (Application) | Isolation of Pathogenic Staphylococci |
| Incubation Conditions | 30°C to 35°C |
| Incubation Time | 24 to 48 to 72 hr. |
| pH | 7.4 ±0.2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?