missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals

Product Code. p-3547201 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11401921

Brand: MP Biomedicals 0219470280

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Physical Form Crystalline
Quantity 100 g
Formula Weight 169
Chemical Name or Material Manganese Sulfate Monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.