missing translation for 'onlineSavingsMsg'
Learn More

Maltopentaose, MedChemExpress

Product Code. 16451648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451648 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451648 Supplier Medchem Express Supplier No. HYN1495

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Maltopentaose is the shortest chain oligosaccharide that can be classified as maltodextrin and is also used in a study to investigate glycation and phosphorylation of α-lactalbumin.

Maltopentaose is the shortest chain oligosaccharide that can be classified as maltodextrin and is also used in a study to investigate glycation and phosphorylation of α-lactalbumin.

Specifications

Chemical Name or Material Maltopentaose
CAS 34620-76-3
Color White
Purity Grade Notes Research
Molecular Formula C30H52O26
Quantity 5 mg
Synonym Maltopentose
Solubility Information H2O : 125 mg/mL (150.84 mM; Need ultrasonic)
SMILES O[C@H]1[C@@H]([C@H](O[C@H](O[C@]([C@@H]2O)([H])[C@H](O[C@H](O[C@@]([C@H](O)CO)([H])[C@H](O)[C@@H](O)C=O)[C@@H]2O)CO)[C@@H]1O)CO)O[C@@]([C@@H]([C@@H](O)[C@@H]3O[C@@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)O)([H])O[C@@H]3CO
Molecular Weight (g/mol) 828.72
Formula Weight 828.72
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.