Learn More
Value Plastics Male Luer Slip to 200 Series Barb
Shop All Cole Parmer Products
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/32 in.
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
Description
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of flexible tubing. Slip luer fittings allow for a simple yet secure connection
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 1/8 in. |
| Inlet | Male Luer Slip, Hose Barb |
| Material (Seal) | Polypropylene |