Learn More
Value Plastics Male Luer Integral Lock Ring to NPT(M) Thread Adapters
Product Code. 30053312 Shop All Cole Parmer Products
Change view
Click to view available options
Thread Size:
1/4-18 NPT(M)
1/8-27 NPT(M)
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size Material (Seal) unitSize
30053312 1/4-18 NPT(M) Nylon Pack of 25
30053162 1/8-27 NPT(M) Polypropylene Pack of 25
30053634 1/4-18 NPT(M) Polypropylene Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30053312 Supplier Value Plastics Supplier No. 5011993
Only null left
Add to basket
Add to basket
Prevent leaks with integral lock ring mechanism
Easily transition from a luer to thread fitting. Luer fittings allow for a simple yet secure twist-lock connection. NPT thread fittings are designed to create a tight seal under pressure
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Thread Size 1/4-18 NPT(M)
Material (Seal) Nylon
Product Type Male Luer Integral Lock Ring Hex