Learn More
Integra Biosciences™ Mains Adapter
Product Code. p-8010700
Click to view available options
Type:
Mains Adapter; EU version
Mains Adapter; UK version
Unit Size:
Each
Product Code. 11686580
Brand: Integra Biosciences™ 156631
Only null left
Add to basket
Add to basket
Use this adapter with Integra Biosciences™ PipetBoy™ Pro Pipettors.
  • Mains Adapter for PIPETBOY pro
  • Input: 100–240 VAC, 50/60 Hz
PIPETEBOY pro delivers precise and efficient serological pipetting
  • Integrated LED providing an optimal illumination of the area around the pipette for accurate pipetting and preventing eyestrain
  • Accurately displace liquid in the pipette by applying variable finger pressure on the pipettor triggers
  • Light as a feather, 190 grams
  • Five colors available
  • Speed can be set continuously via the thumb wheel
  • Red LED indicates when the battery needs to be charged
  • Ergonomic design, following the recommendations of hand shape ergonomics
Specifications
Type Mains Adapter; EU version
For Use With (Equipment) PipetBoy Pro