Learn More
3M™ Magnifying Lens
Click to view available options
Power Intensity:
2.0X
2.5X
Unit Size:
Each
Description
- On the 3M™ Speedglas™ Welding Helmet Series 9100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure
- For the new 3M™ Speedglas™ Welding Helmet Series 100 there is a magnifying lens holder built into the welding filter, making lens installation easier and more secure. For the older 100 series filters you will need a holder for the magnifier
- For the 3M™ Speedglas™ Welding Helmet SL you will need to add the holder for magnifier as a separate spare part (171031)
Specifications
Specifications
| For Use With (Equipment) | Speedglas |
| Power Intensity | 2.0X |
| Product Type | Magnifying Lens |