Learn More
Magnesium carbonate basic, heavy, extra pure
Product Code. 15234734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15234734 10 kg Plastic drum 10kg
10177180 500 g Plastic bottle 500g
15224734 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15234734 Supplier Thermo Scientific Acros Supplier No. 413400100
Request Bulk or Custom Format
Magnesium carbonate basic, heavy, 40 to 45% (MgO), CMgO3, CAS Number-39409-82-0, 500g, loss on calcination: 55 to 60%, White, 208-915-9, 84.31, Extra Pure, MgCO3, 15,5724, MFCD00011107, Plastic bottle, Powder, OM2470000 | CAS: 39409-82-0 | C4H10Mg5O18 | 467.63 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39409-82-0
Molecular Formula C4H10Mg5O18
Molecular Weight (g/mol) 467.63
MDL Number MFCD00149783
InChI Key YMKSTRLFWJQKFP-UHFFFAOYSA-D
IUPAC Name pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
SMILES O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Specifications
Color White
Physical Form Powder
Quantity 10 kg
Linear Formula MgCO3
Merck Index 15,5724
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, soluble in acids
Formula Weight 84.31
Percent Purity 40 to 45% (MgO)
Grade Extra Pure
Packaging Plastic drum
Chemical Name or Material Magnesium carbonate basic, heavy