Learn More
Applied Biosystems™ MagMAX™ Microbiome Lysis Solution
Product Code. 16356582
Change view
Click to view available options
Quantity:
800 mL
Unit Size:
1000 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16356582 800 mL 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16356582 Supplier Applied Biosystems™ Supplier No. A42361
Edge
This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit.
This lysis buffer is included in the MagMAX Microbiome Ultra Nucleic Acid Isolation Kit (Cat. Nos. A42357, and A42358). It is made available separately for applications that require more lysis solution than is provided in the kit.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
Sample Type Bacteria, Blood, Buccal Samples, Cells, Fungi, Liquid Samples (e.g. Serum), Viral Samples, Yeast
For Use With (Application) Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput Compatibility High-throughput Compatible, Manual Protocols, Automated Protocols
Product Line MagMAX
Quantity 800 mL
Shipping Condition Room Temperature
Target Viral DNA⁄RNA, Pathogen DNA⁄RNA, Total Nucleic Acids