Learn More
R&D Systems™ MagCellect Plus Human EpCAM+ Cell Isolation Kit
Description
Epithelial Cellular Adhesion Molecule (EpCAM), also known as KS1/4, gp40, GA733-2, 17-1A, and TROP-1, is a transmembrane glycoprotein that functions as a homophilic Ca2+-independent adhesion molecule. EpCAM is expressed in fetal lung, kidney, liver, pancreas, skin, and germ cells, and in adult epithelia. EpCAM expression is elevated in human malignant neoplasia, with most carcinomas expressing EpCAM.
Specifications
Specifications
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Detection Method | ELISA |
| Quantity | 1 Kit |
| Type | Cell Isolation Kit |
For Research Use Only. Not for use in Humans.