missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521791
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, Molt-4, GL261, BCL-1, Raw264.7, PC-12. Predicted reactivity: Mouse (97%), Rat (97%), Xenopus laevis (86%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is similar to Maf1, a Saccharomyces cerevisiae protein highly conserved in eukaryotic cells. Yeast Maf1 is a negative effector of RNA polymerase III (Pol III). It responds to changes in the cellular environment and represses pol III transcription. Biochemical studies identified the initiation factor TFIIIB as a target for Maf1-dependent repression.
Specifications
| MAF1 | |
| Polyclonal | |
| Unconjugated | |
| MAF1 | |
| 1110068E11Rik; AU042856; homolog of yeast MAF1; MAF1; MAF1 homolog (S. cerevisiae); MAF1 homolog, negative regulator of RNA polymerase III; MAF1 negative regulator of RNA polymerase III; repressor of RNA polymerase III transcription MAF1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84232 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% Thimerosal; pH 7 | |
| Q9H063 | |
| MAF1 | |
| Recombinant protein encompassing a sequence within the center region of human MAF1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction