missing translation for 'onlineSavingsMsg'
Learn More

Macropa-NH2 hydrochloride, MedChemExpress

Product Code. p-200095897
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16494538

Brand: Medchem Express HY111895A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Macropa-NH2 hydrochloride is the precursor of Macropa-NCS. Macropa-NCS is conjugated to Anti-Human HER2 (HY-P9907) as well as to the prostate-specific membrane antigen-targeting compound RPS-070 and is a promising therapeutic radionuclide applied in the treatment of soft-tissue metastases.

Macropa-NH2 hydrochloride is the precursor of Macropa-NCS. Macropa-NCS is conjugated to Anti-Human HER2 (HY-P9907) as well as to the prostate-specific membrane antigen-targeting compound RPS-070 and is a promising therapeutic radionuclide applied in the treatment of soft-tissue metastases.

Specifications

Chemical Name or Material Macropa-NH2 hydrochloride
CAS 2443966-86-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H38ClN5O8
Quantity 10 mg
Solubility Information DMSO : 83.33 mg/mL (142.67 mM; Need ultrasonic)
SMILES O=C(O)C1=NC(CN2CCOCCOCCN(CC3=NC(C(O)=O)=CC=C3)CCOCCOCC2)=CC(N)=C1.[H]Cl
Molecular Weight (g/mol) 584.06
Formula Weight 584.06
Percent Purity 95.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.