Learn More
Thermo Scientific™ MacConkey Agar without Salt
Product Code. 10464203
Change view
Click to view available options
Fill Volume:
17 mL
Fill Weight:
17g ± 5%
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Volume Fill Weight unitSize
10464203 17 mL 17g ± 5% Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10464203 Supplier Thermo Scientific™ Supplier No. PO5131A
Detect and enumerate coliforms in urine samples to diagnose UTIs on this differential medium which prevents Proteus swarming.
Detect and enumerate coliform organisms in clinical samples with Thermo Scientific™ MacConkey Agar without Salt. The differential medium suppresses swarming of Proteus species and is, therefore, recommended for urine examination in the investigation of suspected urinary tract infection (UTI).
MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.
TRUSTED_SUSTAINABILITY
Specifications
Color Salmon pink, transparent
Content And Storage 2° to 12°C, in the dark
Description MacConkey Agar without Salt
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Volume 17 mL
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
For In Vitro Diagnostic Use.