Learn More
M443, MedChemExpress
Product Code. 16484608
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484608 10 mg 10mg
16480907 50 mg 50mg
16455456 100 mg 100mg
16437327 5 mg 5mg
16413166 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16484608 Supplier Medchem Express Supplier No. HY112274
MedChemExpress M443 is an irreversible and specific inhibitor of MRK, with an IC50<125 nM.
M443 is an irreversible and specific inhibitor of MRK, with an IC50<125 nM.
Specifications
Chemical Name or Material M443
CAS 1820684-31-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H30F3N7O2
Quantity 10 mg
Solubility Information DMSO : 55 mg/mL (93.28 mM; Need ultrasonic)
SMILES FC(F)(F)C1=CC(NC(C2=CC(NC3=NC=CC(C4CN(C(C=C)=O)CCC4)=N3)=C(C)C=C2)=O)=CC(N5C=NC(C)=C5)=C1
Molecular Weight (g/mol) 589.61
Formula Weight 589.61
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation