missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M2I-1, MedChemExpress
Description
M2I-1 is a Mad2 inhibitor targeting the binding of Mad2 to Cdc20, an essential protein-protein interaction (PPI) within the spindle assembly checkpoint (SAC).
Specifications
Specifications
| Chemical Name or Material | M2I-1 |
| Physical Form | Solid |
| CAS | 312271-03-7 |
| Color | Orange |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C19H24N4O4S |
| Solubility Information | DMSO : ≥ 32 mg/mL (79.11 mM) |
| SMILES | O=C(/C(C(N1)=O)=C\C2=CC=C(N(CC(C)C)CC(C)C)C([N+]([O-])=O)=C2)NC1=S |
| Molecular Weight (g/mol) | 404.48 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?