missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M13 Phage coat protein g3p Rabbit anti-Virus, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA581762
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for The antibody binds specifically to phage coat protein g3p of fd phage or M13 phage.
M13 antibodies have numerous uses in screening phage display libraries, especially the screening of single chain antibody libraries. The display of repertoires of antibody fragments on the surface of filamentous phage offers a new way to produce immunoreagents with defined specificities. Phage derived antibody fragments offer a number of advantages over mouse monoclonal antibodies, such as better clearance from the blood, the possibility to select from human combinatorial libraries and the relative ease by which such fragments can be manipulated. The phage display technique thus facilitates the selection of antibody fragments of therapeutic value or research interest. Antibodies to M13 filamentous phage coat proteins are instrumental in the selection and detection of phages expressing specific antibody fragments or peptide sequences at their surface.
Specifications
| M13 Phage coat protein g3p | |
| Polyclonal | |
| PBS with no preservative; pH 6 | |
| M13 Bacteriophage coat protein G3P (YP_009111293.1, Ala19-Ala130) | |
| 100 μL | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction