Learn More
Invitrogen™ Lysing Solution, Whole Blood Lysing Solution
Product Code. 10292873
Click to view available options
Volume (Metric):
25 mL
100 mL
Unit Size:
Each
Product Code. 10292873
Brand: Invitrogen™ GAS010S100
Lysing Solution, Whole Blood Lysing Solution
Invitrogen Cal-Lyse™ is a lysing solution formulated to enable the lysis of erythrocytes in samples of anticoagulated human peripheral blood. Invitrogen Cal-Lyse™ lysing solution is intended to be used as an aid in the enumeration of leukocytes that have been stained with monoclonal antibodies for flow cytometric analysis.
View a selection guide for all cell lysis and preservation solutions and products related to sample fixation and permeablization, blocking reagents, and information about the use of Dynabeads™ for cell isolation.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store at room temperature.
  • Shelf life: 24 months from date of manufacture
  • Quantity 100 mL
    Volume (Metric) 100 mL
    Reagent Type Cell Lysis Buffer
    Shipping Condition Room Temperature
    Product Type Whole Blood Lysing Solution