missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Lysine Decarboxylase Broth (Taylor)
Description
Detect lysine decarboxylase enzyme activity in by salmonellae and other Enterobacteriaceae with Lysine Decarboxylase Broth (Taylor), which helps differentiate enteric bacteria based on their ability to decarboxylate lysine and produce cadaverine.
Specifications
Specifications
| Color | Pearl Violet, Transparent |
| Content And Storage | 2°C to 12°C |
| Format | Poured Tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Fill Weight | 5.5 to 6.5 g |
| Packaging Type | 50 Tubes/Box |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?