missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYPLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
206.00 EUR - 483.00 EUR
Specifications
| Antigen | LYPLA2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18362467
|
Novus Biologicals
NBP3-04729-20UL |
20 μL |
206.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18355776
|
Novus Biologicals
NBP3-04729-100UL |
100 μL |
483.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYPLA2 Polyclonal specifically detects LYPLA2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| LYPLA2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 11313 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| acyl-protein thioesterase 2, APT2, DJ886K2.4, EC 3.1.2, EC 3.1.2.-, LPL-II, lysophospholipase IIAPT-2LysoPLA II | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 147-231 of human LYPLA2 (NP_009191.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title