missing translation for 'onlineSavingsMsg'
Learn More

LYN-1604 hydrochloride, MedChemExpress

Product Code. 16493638
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16493638

Brand: Medchem Express HY101923A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress LYN-1604 hydrochloride is a potent UNC-51-like kinase 1 (ULK1) activator (EC50=18.94 nM) for the research of triple negative breast cancer (TNBC).

LYN-1604 hydrochloride is a potent UNC-51-like kinase 1 (ULK1) activator (EC50=18.94 nM) for the research of triple negative breast cancer (TNBC).

Specifications

Chemical Name or Material LYN-1604 hydrochloride
CAS 2216753-86-3
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C33H44Cl3N3O2
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (161.01 mM; Need ultrasonic)
SMILES ClC1=CC=C(C(CN2CCN(C(CN(CC(C)C)CC(C)C)=O)CC2)OCC3=CC=C(C=CC=C4)C4=C3)C(Cl)=C1.[H]Cl
Molecular Weight (g/mol) 621.08
Formula Weight 621.08
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.