missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYN-1604 hydrochloride, MedChemExpress
Description
LYN-1604 hydrochloride is a potent UNC-51-like kinase 1 (ULK1) activator (EC50=18.94 nM) for the research of triple negative breast cancer (TNBC).
Specifications
Specifications
| Chemical Name or Material | LYN-1604 hydrochloride |
| CAS | 2216753-86-3 |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C33H44Cl3N3O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 100 mg/mL (161.01 mM; Need ultrasonic) |
| SMILES | ClC1=CC=C(C(CN2CCN(C(CN(CC(C)C)CC(C)C)=O)CC2)OCC3=CC=C(C=CC=C4)C4=C3)C(Cl)=C1.[H]Cl |
| Molecular Weight (g/mol) | 621.08 |
| Formula Weight | 621.08 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?