missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LYK5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17208763 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17208763 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17208763 Supplier Invitrogen™ Supplier No. PA5115978

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total STRAD.

LYK5; STE20-related Adaptor Protein (STRAD); FLJ90524. Gene Map Locus is 17q23.3 and members of STE-20 like kinase family are known to stimulate MAPK pathways by directly activating MAPKKK. STRAD is a novel pseudokinase member of this family consisting of a STE-20 like kinase domain but lacks several residues that are required for its catalytic activity. It specifically binds LKB1 and plays a key role in regulating tumor suppressor activities of LKB1. It functions as an upstream activator of LKB1 and also directs the sub-cellular localization of LKB1 by anchoring it in the cytoplasm. STRAD-LKB1 interaction results in phosphorylation of STRAD and enhanced autophosphorylation of LKB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LYK5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene STRADA
Gene Accession No. Q3UUJ4, Q7RTN6
Gene Alias 2610019A05Rik; 6030402H20Rik; AI480680; E130112C09Rik; LYK5; NY-BR-96; PMSE; protein kinase LYK5; serologically defined breast cancer antigen NY-BR-96; STE20-like pseudokinase; STE20-related adapter protein; STE20-related kinase adapter protein alpha; STE20-related kinase adaptor alpha; Stlk; Strad; STRAD alpha; Strada
Gene Symbols STRADA
Host Species Rabbit
Immunogen A synthesized peptide derived from human STRADA(Accession Q7RTN6), corresponding to amino acid residues M312-R362.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72149, 92335
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.