missing translation for 'onlineSavingsMsg'
Learn More

LY404039, MedChemExpress

Product Code. p-200096201
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16478138

Brand: Medchem Express HY50906

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY404039 is a potent, selective and orally active mGluR2 and mGluR3 agonist with Kis of 149 nM and 92 nM for recombinant human mGluR2 and mGluR3, respectively. LY404039 shows >100-fold selectivity for mGluR2/3 over other receptors/transproters. LY404039 has antipsychotic and anxiolytic effects.

LY404039 is a potent, selective and orally active mGluR2 and mGluR3 agonist with Kis of 149 nM and 92 nM for recombinant human mGluR2 and mGluR3, respectively. LY404039 shows >100-fold selectivity for mGluR2/3 over other receptors/transproters. LY404039 has antipsychotic and anxiolytic effects.

Specifications

Chemical Name or Material LY404039
Physical Form Solid
CAS 635318-11-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C7H9NO6S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 5.2 mg/mL (22.11 mM; ultrasonic and adjust pH to 6 with NaOH) ∣DMSO : 3.84 mg/mL (16.33 mM; Need ultrasonic and warming)
SMILES O=C([C@]1(N)CS([C@@]2([H])[C@H](C(O)=O)[C@@]12[H])(=O)=O)O
Molecular Weight (g/mol) 235.21
Formula Weight 235.21
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.