Learn More
LY341495, MedChemExpress
Product Code. 16440628
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Product Code. 16440628
Brand: Medchem Express HY70059
MedChemExpress LY341495 is a metabotropic glutamate receptor (mGluR) antagonist with IC50s of 21 nM, 14 nM, 7.8 μM, 8.2 μM, 170 nM, 990 nM, 22 μM for mGlu2, mGlu3, mGlu1a, mGlu5a, mGlu8, mGlu7, and mGlu4 receptors, respectively.
LY341495 is a metabotropic glutamate receptor (mGluR) antagonist with IC50s of 21 nM, 14 nM, 7.8 μM, 8.2 μM, 170 nM, 990 nM, 22 μM for mGlu2, mGlu3, mGlu1a, mGlu5a, mGlu8, mGlu7, and mGlu4 receptors, respectively.
Specifications
Chemical Name or Material LY341495
CAS 201943-63-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H19NO5
Quantity 5 mg
Solubility Information DMSO : 6 mg/mL (16.98 mM; Need ultrasonic)
SMILES O=C(O)[C@@H]1[C@H](C1)[C@@](N)(CC2C3=C(C=CC=C3)OC4=C2C=CC=C4)C(O)=O
Molecular Weight (g/mol) 353.37
Formula Weight 353.37
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation