Learn More
LY3295668, MedChemExpress
Product Code. 16467547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467547 5 mg 5mg
16461067 50 mg 50mg
16405606 100 mg 100mg
16404808 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16467547 Supplier Medchem Express Supplier No. HY114258
MedChemExpress LY3295668 (AK-01) is a potent, orally active and highly specific Aurora-A kinase inhibitor, with Ki values of 0.8 nM and 1038 nM for AurA and AurB, respectively.
LY3295668 (AK-01) is a potent, orally active and highly specific Aurora-A kinase inhibitor, with Ki values of 0.8 nM and 1038 nM for AurA and AurB, respectively.
Specifications
Chemical Name or Material LY3295668
CAS 1919888-06-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H26ClF2N5O2
Quantity 5 mg
Synonym AK-01
Solubility Information DMSO : 100 mg/mL (204.10 mM; Need ultrasonic)
SMILES O=C([C@@]1(CC2=NC(NC3=NNC(C)=C3)=CC=C2F)C[C@@H](C)N(CC4=CC=CC(Cl)=C4F)CC1)O
Molecular Weight (g/mol) 489.95
Formula Weight 489.95
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease