Learn More
LY2857785, MedChemExpress
Product Code. 16475806
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
Product Code. 16475806
Brand: Medchem Express HY12293
MedChemExpress LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9 (IC50 11 nM) and other transcription kinases CDK8 (IC50 16 nM), and CDK7 (IC50 246 nM).
LY2857785 is a type I reversible and competitive ATP kinase inhibitor against CDK9 (IC50 11 nM) and other transcription kinases CDK8 (IC50 16 nM), and CDK7 (IC50 246 nM).
Specifications
Chemical Name or Material LY2857785
CAS 1619903-54-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H36N6O
Quantity 100 mg
Solubility Information DMSO : 10 mg/mL (22.29 mM; Need ultrasonic)
SMILES CC(C1=C2C=C(C3=NC(N[C@H]4CC[C@H](NC5CCOCC5)CC4)=NC=C3)C=CC2=NN1C)C
Molecular Weight (g/mol) 448.6
Formula Weight 448.6
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease