Learn More
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-5931-38
Description
The RB6-8C5 MAb reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets.
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, Functional Assay
Specifications
| Ly-6G/Ly-6C | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.2 | |
| P0CW02, P0CW03, P35461 | |
| Rat | |
| 5 mg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
| Flow Cytometry, Functional Assay | |
| RB6-8C5 | |
| Functional Grade | |
| Ly6c2 | |
| AA682074; AA959465; Gr1; Gr-1; Granulocyte Marker; Ly6c; Ly-6C; Ly-6C.2; Ly6c1; Ly-6C1; Ly6c2; Ly-6C2; Ly6g; Ly-6G; ly-6G.1; lymphocyte antigen 6 complex, locus C; lymphocyte antigen 6 complex, locus C1; lymphocyte antigen 6 complex, locus C2; lymphocyte antigen 6 complex, locus G; lymphocyte antigen 6C precursor (Ly-6C); lymphocyte antigen 6C1; Lymphocyte antigen 6C2; lymphocyte antigen 6G; Lymphocyte antigen Ly-6C; lymphocyte differentiation antigen; PML; PMN | |
| Affinity chromatography | |
| RUO | |
| 100041546, 17067, 546644 | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.