Learn More
Tocris Bioscience™ LY 2389575 hydrochloride
Selective negative allosteric modulator of mGlu3
Brand:  Tocris Bioscience™ 4969/10
Encompass
Product Code. 15766356
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
LY 2389575 hydrochloride is a selective negative allosteric modulator of mGlu3 (IC50 = 190 nM). Exhibits > 65-fold selectivity for mGlu3 over other mGlu receptors. Abolishes the neuroprotective action of LY 379268 against amyloid β toxicity in mixed cortical neuronal and astrocyte cell cultures.
Specifications
Specifications
>98%
10 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions