Learn More
Welch™ LVS 101 Z Laboratory Vacuum System
Product Code. 11849081 Shop All Welch Ilmvac Products
Change view
Click to view available options
Depth (Metric):
310 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
11849081 310 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11849081 Supplier Welch™ Supplier No. 115027
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Modular manual and automatic control chemically resistant PTFE construction
Modular manual and automatic control chemically resistant PTFE construction 230V 50Hz flow rate 17L/min vacuum mbar <8 Ilmvac
TRUSTED_SUSTAINABILITY
Specifications
Electrical Requirements 230 V 50/60 Hz
Depth (Metric) 310 mm
Model LVS 101 Z
Frequency 50/60 Hz
Overall Height (Metric) 445 mm
Overall Width (Metric) 360 mm
Pump Type Laboratory Vacuum System
Maximum Pressure <8 mbar
Voltage 230 V