Learn More
Value Plastics Luer Thread Fittings, Elbow
Product Code. 30053834 Shop All Cole Parmer Products
Change view
Click to view available options
Inlet:
Female Luer Thread
Male Luer to Female Luer
Male Luer, Female Luer Thread
Material (Seal):
Nylon
Polypropylene
Unit Size:
Pack of 25
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Inlet Material (Seal) unitSize
30053834 Male Luer, Female Luer Thread Nylon Pack of 25
30054208 Female Luer Thread Polypropylene Pack of 25
30053705 Male Luer to Female Luer Polypropylene Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30053834 Supplier Value Plastics Supplier No. 5012007
Only null left
Add to basket
Add to basket
Easily transition to a luer fitting with directional flexibility
Quick and easy assembly and disassembly without the need for tools. Luer fittings allow for a simple yet secure connection. Conforms to ISO-594-1 and ISO 594-2 standards
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;RoHS
Inlet Male Luer, Female Luer Thread
Material (Seal) Nylon