missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lubricating Oil Calibration Set (contains Ba, Ca, Cl, Mg, Mo, P, S, Si, Zn at randomized concentrations), VHG™
Description
Lubricating Oil Calibration Set (contains Ba, Ca, Cl, Mg, Mo, P, S, Si, Zn at randomized concentrations)
Specifications
Specifications
| Chemical Name or Material | Lube Oil Calibration Set |
| Methods | XRF |
| Analyte or Component Names | Lube Oil Calibration Set |
| Shipping Condition | Room Temperature |
| IUPAC Name | Lube Oil Calibration Set |
| Solvent or Matrix | Lubricating Oil |
| Quantity | 23 x 50 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?