missing translation for 'onlineSavingsMsg'
Learn More

Lubiprostone Impurity 2, TRC

Product Code. 30469397
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469397 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469397 Supplier Toronto Research Chemicals Supplier No. TRCL4735185MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lubiprostone Impurity 2
CAS 2196182-47-3
Molecular Formula C20 H30 F2 O4
Quantity 5 mg
Synonym 7-((1R,2S)-2-(4,4-Difluoro-3-oxooctyl)-5-oxocyclopent-3-en-1-yl)heptanoic Acid
InChI Formula InChI=1S/C20H30F2O4/c1-2-3-14-20(21,22)18(24)13-11-15-10-12-17(23)16(15)8-6-4-5-7-9-19(25)26/h10,12,15-16H,2-9,11,13-14H2,1H3,(H,25,26)/t15-,16-/m1/s1
SMILES CCCCC(F)(F)C(=O)CC[C@H]1C=CC(=O)[C@@H]1CCCCCCC(=O)O
IUPAC Name 7-[(1R,2S)-2-(4,4-difluoro-3-oxooctyl)-5-oxocyclopent-3-en-1-yl]heptanoic acid
Molecular Weight (g/mol) 372.447
Formula Weight 372.211
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.