Learn More
Schuett Biotec™ EcoVac Vacuum Pump
Description
- Autoclavable safety-bottle
- Low-noise and low-vibration double chambered membrane pump
- Excellent cost-efficiency
- Optional double chambered membrane/vacuum pump
- Option foot-switch
Specifications
Specifications
| Max. Vacuum | 300 mbar |
| Electrical Requirements | 230 V |
| Dimensions (L x W x H) | 80 x 60 x 160 mm |
| Model | LD90 |
| Weight (Metric) | ∼850 g |
| Flow Rate | 4 L/min. |
| Frequency | 50/60 Hz |
| Pump Type | Vacuum Pump |
| Voltage | 230 V |