Learn More
Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Bath
Saves up to 80% against standard compressor units. Grant Instruments™ ecocool™ Refrigerated/Heated Circulating Baths are supplied assembled as ready to use kits, complete with accessory hosing, clips and connectors as standard.
Brand: Grant Instruments™ ECOCOOL_100R
This item is not returnable.
View return policy
Description
- Eco mode optimises the cooling control needed, enabling rapid cool down when required.
- True energy saving against competitor units that only switch the compressor on or off.
- Intuitive user interface
- No-spill valved front drain.
- Built in Britain to the highest specifications, 5 day dispatch, 4 year industry leading warranty.
- Modern, sleek, attractive design
- Thermostat and chiller work in harmony, neither will operate alone, eliminating any danger of overheating or freezing.
- Single front switch for user convenience.
- No side vents - locate to suit the user, not the unit.
Specifications
| 5 L | |
| -20°C to +100°C | |
| ecocool™ 100 | |
| 0.05°C | |
| 245 mm | |
| 640 mm | |
| 29 kg | |
| 50/60 Hz |
| 250W at 20°C, 230W at 10°C, 185W at 0°C, 115W at -10°C, 40W at -10°C | |
| 4 digit LED | |
| 3 | |
| 4 Years | |
| 154 mm | |
| 118 mm | |
| 120/230 V |