Learn More
Corning™ LSE™ Shaking Incubator Accessory, Rack Holders
Product Code. 16875721 Shop All Corning Products
Change view
Click to view available options
Holds:
15 mL Test Tubes
50 mL Test Tubes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
16875721 50 mL Test Tubes Each
16865721 15 mL Test Tubes -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16875721 Supplier Corning™ Supplier No. 480169
For use with LSE 71L Shaking Incubator
Note: These products are for research use only. Not intended for use in diagnostic or therapeutic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Holds 50 mL Test Tubes
Product Type Rack Holder
For Use With (Equipment) LSE Shaking Incubator, 71L