missing translation for 'onlineSavingsMsg'
Learn More

LPCAT1 Mouse anti-Human, Mouse, Rat, Clone: 8B6E9, Proteintech

Product Code. 16847565 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16847565 20 μL 20µL
16837565 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16847565 Supplier Proteintech Supplier No. 660441IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lysophosphatidylcholine (LPC) acyltransferase (LPCAT; EC 2.3.1.23) catalyzes the conversion of LPC to phosphatidylcholine (PC) in the remodeling pathway of PC biosynthesis (Nakanishi et al., 2006

Specifications

Antigen LPCAT1
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone 8B6E9
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LPCAT1
Gene Accession No. Q1HAQ0, Q3TFD2, Q8NF37
Gene Alias Acyltransferase like 2, AYTL2, LPC acyltransferase 1, lpcat, LPCAT 1, LPCAT1, Lyso PAF acetyltransferase, LysoPAFAT, LysoPC acyltransferase 1, PFAAP3
Gene Symbols Lpcat1
Host Species Mouse
Immunogen LPCAT1 Fusion Protein Ag9060
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210992, 361467, 79888
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.