Learn More
LP-211, MedChemExpress
Product Code. 16474458
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474458 10 mg 10mg
16443096 25 mg 25mg
16417187 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16474458 Supplier Medchem Express Supplier No. HY111455
MedChemExpress LP-211 is a selective and blood−brain barrier penetrant 5-HT7 receptor agonist, with a Ki of 0.58 nM, with high selectivity over 5-HT1A receptor (Ki, 188 nM) and D2 receptor (Ki, 142 nM).
LP-211 is a selective and blood−brain barrier penetrant 5-HT7 receptor agonist, with a Ki of 0.58 nM, with high selectivity over 5-HT1A receptor (Ki, 188 nM) and D2 receptor (Ki, 142 nM).
Specifications
Chemical Name or Material LP-211
Physical Form Oil
CAS 1052147-86-0
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C30H34N4O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (214.31 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (107.15 mM; Need ultrasonic)
SMILES O=C(NCC1=CC=C(C#N)C=C1)CCCCCN2CCN(C3=CC=CC=C3C4=CC=CC=C4)CC2
Molecular Weight (g/mol) 466.62
Formula Weight 466.62
Percent Purity 99.55%
Grade Research