missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LOXL3 Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Product Code. 17950233
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17950233 200 μL 200µL
17919122 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17950233 Supplier Invitrogen™ Supplier No. PA5145269

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LOXL3 is a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOXL3
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene LOXL3
Gene Accession No. P58215, Q9Z175
Gene Alias Lor2; LOW QUALITY PROTEIN: lysyl oxidase homolog 3; LOXL; Loxl2; Loxl3; Lysyl oxidase homolog 3; lysyl oxidase like 3; lysyl oxidase-like 2; lysyl oxidase-like 3; lysyl oxidase-like protein 3; Lysyl oxidase-related protein 2
Gene Symbols LOXL3
Host Species Rabbit
Immunogen Recombinant Human LOXL3 protein, Thr519-Asn726 (Accession #P58215).
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100512652, 16950, 312478, 84695
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.