Learn More
Thermo Scientific Chemicals Low Form Nickel Crucible;Cap (ml), 35;Top Dia (mm), 46;Bottom Dia (mm), 38;Depth (mm), 30
Product Code. 15403605
Click to view available options
Capacity (Metric):
15 mL
20 mL
25 mL
35 mL
55 mL
100 mL
250 mL
Diameter (Metric) Outer:
33mm
36mm
45mm
46mm
47mm
59mm
82mm
Height (Metric):
23mm
30mm
35mm
43mm
46mm
60mm
Unit Size:
Each
Product Code. 15403605
Brand: Thermo Scientific Chemicals 035948.KT
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Low Form
Material Nickel
Capacity (Metric) 35 mL
Diameter (Metric) Outer 46mm
Height (Metric) 30mm
Product Type Nickel Crucible
Diameter (Metric) Base 38mm