Learn More
Lovibond™ Phenol Red Tablet
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Description
Lovibond Phenol Red Tablet, Orange, Composition: Sodium Chloride, Format: Tablet, For Use With: Water analysis, Colorimeter and spectrophotometer, Method: Phenol red, pH Range: 6.5 to 8.4, Shelf Life: 10 Years, Storage Requirements: 20 deg.C
Specifications
Specifications
| Color | Orange |
| Composition | Sodium Chloride |
| pH Range | 6.5 to 8.4 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | Colorimeter and spectrophotometer |
| Quantity | 100/Pk. |
| Storage Requirements | 20°C |
| Test Method | Phenol red |
| Type | Phenol Red Tablet |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur