Learn More
Lovibond™ Aqua Comparitor Color Discs: Silica
For testing water with the Aqua Comparator
Brand: Lovibond™ 146350
Additional Details : Weight : 0.02900kg
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for silica using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ | |
0.25 to 4mg/L | |
Silica LR Color Disc |