missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Aqua Comparitor Color Discs: Silica
For testing water with the Aqua Comparator
26.10 EUR
Specifications
For Use With (Application) | Testing water with the Aqua comparator™ |
---|---|
For Use With (Equipment) | Aqua comparator™ |
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | ||||
11705558
|
Lovibond™
146360 |
Silica VLR Color Disc | 0 to 1mg/L |
26.10 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
11785548
|
Lovibond™
146350 |
Silica LR Color Disc | 0.25 to 4mg/L |
26.10 EUR
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
Description
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for silica using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Testing water with the Aqua comparator™ | |
Aqua comparator™ |