Learn More
Lotilaner, MedChemExpress
Product Code. 16494858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16494858
Brand: Medchem Express HY116564
MedChemExpress Lotilaner is a parasiticide, acts as a potent non-competitive antagonist of insects GABACl receptors, with an IC50 of 23.84 nM for Drosophila melanogaster GABA receptor. No effect on a dog GABAA receptor.
Lotilaner is a parasiticide, acts as a potent non-competitive antagonist of insects GABACl receptors, with an IC50 of 23.84 nM for Drosophila melanogaster GABA receptor. No effect on a dog GABAA receptor.
Specifications
Chemical Name or Material Lotilaner
Physical Form Solid
CAS 1369852-71-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H14Cl3F6N3O3S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (167.57 mM)
SMILES O=C(NCC(NCC(F)(F)F)=O)C1=C(C=C(S1)C2=NO[C@](C3=CC(Cl)=C(C(Cl)=C3)Cl)(C2)C(F)(F)F)C
Molecular Weight (g/mol) 596.76
Formula Weight 596.76
Percent Purity 99.25%
Grade Research