missing translation for 'onlineSavingsMsg'
Learn More

Lotilaner, MedChemExpress

Product Code. 16455646
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455646

Brand: Medchem Express HY116564

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lotilaner is a parasiticide, acts as a potent non-competitive antagonist of insects GABACl receptors, with an IC50 of 23.84 nM for Drosophila melanogaster GABA receptor. No effect on a dog GABAA receptor.

Lotilaner is a parasiticide, acts as a potent non-competitive antagonist of insects GABACl receptors, with an IC50 of 23.84 nM for Drosophila melanogaster GABA receptor. No effect on a dog GABAA receptor.

Specifications

Chemical Name or Material Lotilaner
Physical Form Solid
CAS 1369852-71-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H14Cl3F6N3O3S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 100 mg/mL (167.57 mM)
SMILES O=C(NCC(NCC(F)(F)F)=O)C1=C(C=C(S1)C2=NO[C@](C3=CC(Cl)=C(C(Cl)=C3)Cl)(C2)C(F)(F)F)C
Molecular Weight (g/mol) 596.76
Formula Weight 596.76
Percent Purity 99.25%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.