Learn More
Thermo Scientific Chemicals Losartan potassium salt
Product Code. 15841052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15841052 25 g 25g
15831052 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15841052 Supplier Thermo Scientific Chemicals Supplier No. 462740250
Request Bulk or Custom Format
CAS: 124750-99-8 | C22H22ClKN6O | 461.01 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 124750-99-8
Molecular Formula C22H22ClKN6O
Molecular Weight (g/mol) 461.01
InChI Key OXCMYAYHXIHQOA-UHFFFAOYSA-N
PubChem CID 53396294
Specifications
Quantity 25 g
Chemical Name or Material Losartan potassium salt