missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide.HCl, 1mg/ml in Methanol (as free base), Lipomed
Description
Loperamide.HCl, 1mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Loperamide Hydrochloride |
| CAS | 34552-83-5 |
| Analyte or Component Names | Loperamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H33 Cl N2 O2 . Cl H |
| Concentration | 1 mg/mL |
| Synonym | 4-(4-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinebutanamide monohydrochloride, 4-(p-Chlorophenyl)-4-hydroxy-N,N-dimethyl-alpha,alpha-diphenyl-1-piperidinobutyramide hydrochloride, Arret, Blox, Brek, Dissenten, Fortasec, Imodium, Imodium A-D, Imosec, Imossel, Lopemid, Lopemin, Loperamide hydrochloride, Loperazine, Loperyl, Lopodium, Maalox Antidiarrheal, NSC 696356, PJ 185, R 18553, Suprasec, Tebloc |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN(C)C(=O)C(CCN1CCC(O)(CC1)c2ccc(Cl)cc2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?