Learn More
SeaKem™ ME Agarose, Lonza™
Product Code. 11597765
Click to view available options
Quantity:
25 g
125 g
500 g
Unit Size:
125g
25g
Each
Product Code. 11597765
Brand: Lonza 50011
  • For serum protein electrophoresis and nucleic acid electrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SeaKem ME Agarose
Quantity 25 g