Learn More
SeaKem™ HGT Agarose, Lonza™
Product Code. 11597775
Change view
Click to view available options
Quantity:
25 g
125 g
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11597775 25 g Each
10407885 125 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11597775 Supplier Lonza Supplier No. 50041
  • Used in counterimmunoelectrophoresis and crossed immunoelectrophoresis
  • High gelling temperature (42°C ± 1.5°C)
  • High gel strength (≥1000g/cm2) and clarity
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material SeaKem HGT Agarose
Quantity 25 g